Nickel

Supplier: VWR International

P50070/77 NN0110-100G NN0100-500G
470149-002CS 1933.9 CAD
470149-002 470301-850 470301-852
Nickel
Chemicals for Science Education

CAS Number: 7440-02-0
Formula Weight: 58.71
Formula: Ni
Density (g/mL): 8.90
Boiling Point (°C): 2732
Freezing Point (°C): 1452
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR