n-Butyric Acid

Supplier: ALDON CORP SE

BB0255-100ML BB0255-500ML
470300-510EA 62.4 CAD
470300-510 470300-512
n-Butyric Acid
Chemicals for Science Education

CAS Number: 107-92-6
Formula Weight: 88.11
Formula: CH3(CH2)2COOH
Hazard Info: Corrosive, Irritant, Strong Odor
Density (g/mL): 0.959
Boiling Point (°C): 163.5
Freezing Point (°C): -5.2
Synonyms: Ethylacetic Acid, Butanoic Acid
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR