n-Butyl Alcohol

Supplier: ALDON CORP SE

BB0235-500MLR BB0235-500ML BB0235-100MLR
470301-512EA 60.6 CAD
470301-512 470301-836 470301-838
n-Butyl Alcohol
Chemicals for Science Education

CAS Number: 71-36-3
Formula: CH3CH23OH
Density: 0.8108 g/mL
Boiling and Freezing Point: 117.5°C, -89°C
Solubility: Water, Miscible with Alcohol and Ether
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a flammable and toxic irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR