Naphthalene

Supplier: ALDON CORP SE

NN0010-100G NN0010-500GF NN0010-500G
470045-578EA 23.55 CAD
470045-578 470301-834 470301-830
Naphthalene
Chemicals for Science Education

CAS Number: 91-20-3
Formula Weight: 128.18
Formula: C10H8
Density (g/mL): 1.145
Boiling Point (°C): 218 Sublimes
Freezing Point (°C): 80
Synonyms: White Tar, Tar Camphor
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR