Ward's® Mitotic Stage Counts Flashcards
Supplier: Ward's
Ward's®
323378
470022-904KT 66.2 CAD
470022-904
Ward's® Mitotic Stage Counts Flashcards
Educational Materials Biology Educational Materials Biology Flashcards
Wide Field of View Shows Many Cells Captured at Various Phases of Mitosis
  • Flashcards are the perfect supplement to any lab that requires the observation of plant mitosis
Use the color photos of plant root tip mitosis to identify and quantify the different stage of mitosis shown on each card and to determine the relative duration for each stage.