Miniature Lamp Sockets
Supplier: VWR International
PH1024-SPL
470110-308PK 44.3 CAD
470110-308
Miniature Lamp Sockets
Electrical Sockets
Sure to Come in Handy.
Why buy a whole new receptacle when you just need to replace the socket? These sockets come complete with terminals for mounting them to the binding holes in universal miniature screw receptacles. Package of 12.