Mineral Oil

Supplier: ALDON CORP SE

MM0590-PK/2 MM0585-500ML MM0585-1L MM0586-500ML
470301-504PK 127.9 CAD
470301-504 470301-505 470301-506 470301-822
Mineral Oil
Chemicals for Science Education

CAS Number: 8042-47-5
Formula Weight: Variable
Formula: CH2[CH2]nCH3
Density (g/mL): 0.818-0.880
Solubility: Organic Solvents
Synonyms: White Mineral Oil, Light Paraffin Oil
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR