Microplates
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
4-421 82.1581
470016-176PK 52.95 CAD
470016-176 470199-952
Microplates
Microplates HTS Microplates
These microplates replace standard test tubes and beakers. The dual-well plates are made of high-quality polystyrene and have 12 large wells and 48 small wells.
  • Closure type: slip-on lid
  • Base shape: flat base
  • Number of wells: 96
  • Max. volume: 390 µl
They are not for use with organic solvents.