Edvotek® Micropipetting Basics
Supplier: Edvotek
S-44
470106-954EA 129.05 CAD
470106-954
Edvotek® Micropipetting Basics
Pipettes Micropipettes
Teach your students how to use a micropipette with ease and accuracy with multicolored dyes. A fun and cost-effective way to learn this important skill.
  • Complete in one class period (45 minutes)
  • Includes instructions, various colored dye samples, and a Pipette Card™
  • All you need is micropipette and tips
Store at room temperature.