Microdissection Forceps
Supplier: Dunrite Instruments
141420 141004
470018-990EA 7.2 CAD
470018-990 470018-954
Microdissection Forceps
Forceps
Deep serrations in the jaws, plus aligning guide pins, make it easier to grasp and hold small objects. Choose from straight or curved points, both with a rounded tip.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.