Microbial Ecology in a Closed System Kit
Supplier: Avantor
WARD470100-972
470100-972KT 495.59 CAD
470100-972
Microbial Ecology in a Closed System Kit
Educational Materials Biology Educational Materials Ecology Learning Activities
Design a Winogradsky Column
  • Demonstrate the use of a Winogradsky column
  • Study population succession
  • Includes a teacher's guide
Study population succession in an ecosystem filled with mud, shredded paper, salts and water. Use this kit to introduce students to the Winogradsky column, a small vertical pond designed to simulate various environmental conditions.
Includes materials for 30 students and a teacher’s guide with background information.