Metric Micrometer
Supplier: WALTER STERN INC
601A
470110-218EA 176.85 CAD
470110-218
Metric Micrometer
Micrometers
Forged steel fork and hammertone finish make ideal for heavy use. Steel frame has satin chrome finished barrel with clearly graduated markings. Includes positive spindle lock and friction stop. Accuracy 0.076mm.
Includes positive spindle lock and friction stop.
Metric scale: 25mm.