Methylene Blue

Supplier: ALDON CORP SE

MM0451-100ML MM0451-500ML MM0440-500ML MM0430-25G MM0450-500ML MM0440-1L MM0440-100ML MM0430-10G
470301-492EA 32.4 CAD
470301-492 470301-493 470301-497 470301-498 470301-804 470301-806 470301-808 470301-814
Methylene Blue
Chemicals for Science Education

CAS Number: 61-73-4
Formula: C16H18N3SCl
Density: 1 g/mL
Boiling Point: Decomposes
Solubility: Water, Alcohol, and Chloroform
Synonyms: Basic Blue 9, Methylene Blue Chloride
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR