Methyl Orange

Supplier: ALDON CORP SE

MM0480-500ML MM0480-100ML MM0465-25GR
470301-484EA 17.85 CAD
470301-484 470301-782 470301-786
Methyl Orange
Chemicals for Science Education

CAS Number: 547-58-0
Formula Weight: 327.35
Formula: C14H14N3NaO3S
Density (g/mL): 0.35-0.39
Freezing Point (°C): >300
Solubility: Hot Water
Synonyms: C.I. No. 13025, C.I. Acid Orange 52
Shelf Life (months): 36
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR