Methyl Cellulose

Supplier: ALDON CORP SE

MM0400-100ML MM0400-500ML MM0380-100G
470301-478EA 19.75 CAD
470301-478 470301-774 470301-776
Methyl Cellulose
Chemicals for Science Education

CAS Number: 9004-67-5
Formula Weight: Mixture
Formula: Mixture
Boiling Point (°C): Decomposes
Solubility: Warm Water
Synonyms: Methocel®, Methyl Ether Cellulose
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR