Metamorphic Rock Set of 18
Supplier: Avantor
WARD470106-754
470106-754KT 60.69 CAD
470106-754
Metamorphic Rock Set of 18
Specimens Rock Specimens Metamorphic Rocks
Includes: six each of marble, slate, and quartzite. Set of 18.
Common rocks and minerals for testing and identification studies are 2–3cm each.