Wards® Metamorphic Rock Collection
Supplier: Avantor
470175-704EA 277.25 CAD
470175-704 470025-226
Wards® Metamorphic Rock Collection
Specimens Rock Specimens Metamorphic Rocks
Perfect for Introductory Lessons on Metamorphic Rocks.
  • Hand cut by Wards® geologists
  • Specimen list with localities included
Study textures, grain sizes, and other features altered by pressure, temperature, and other natural forces, characteristic of metamorphic rocks. The set is housed in a compartmented collection box and comes with an identification list.
Contents:
12 numbered samples, 1×2"
Rocks:
gneiss (2), marble (2), quartzite, schist (3), serpentinite, slate (2), soapstone
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed in individual, labeled plastic display trays and comes with an identification list.
Contents:
12 numbered samples, 3×4"
Caution: Due to the nature of this product it may contain asbestos.