Medium Grade, 15 μm
Supplier: United Scientific Supplies
FPR125 FPR009 FPR011 FPR150 FPR007 FPR240 FPR320
470019-886BX 17.2 CAD
470019-886 470019-882 470019-884 470019-888 470349-724 470349-776 470349-756
Medium Grade, 15 μm
Filters Filter Papers Qualitative Filter Paper
The most widely used qualitative filter paper for routine applications.
  • Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
Packaging: Packages of 100.