Media Bottles
Supplier: DWK Life Sciences (KIMBLE)
219755 219757 219759
470205-564CS 604.14 CAD
470205-564 470205-566 470205-568
Media Bottles
Bottles Glass Bottles
Borosilicate Glass.
  • Large opening, no-drip-lip, and sloping shoulders for easy pouring
  • Approximate volume graduations
  • Large writing area
Both the bottles and rubber-lined caps are autoclavable.