Mayo Scissors
Supplier: Dunrite Instruments
141122 140992
470007-568EA 17.05 CAD
470007-568 470018-948
Mayo Scissors
Scissors
Safe yet sharp.
  • Single blunt side prevents tissue damage
  • Available in 2 lengths
  • Stainless construction resists rust
Designed for use in the famous clinic, these straight-blade scissors allow for precise control. They are offered in two lengths. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.