Magnesium Oxide

Supplier: ALDON CORP SE

MM0120-500G MM0110-100G MM0110-500G
470301-674EA 55.45 CAD
470301-674 470301-676 470301-678
Magnesium Oxide
Chemicals for Science Education

CAS Number: 1309-48-4
Formula Weight: 40.30
Formula: MgO
Hazard Info: Toxic
Density (g/mL): 3.65-3.75
Boiling Point (°C): 3600
Freezing Point (°C): 2800
Solubility: Acids and Slightly in Water
Synonyms: Magnesia, Calcinated Magnesia, Calcined Brucite
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR