Magnesium Hydroxide
Supplier: ALDON CORP SE
CAS Number: 1309-42-8
Formula Weight: 58.32
Formula: Mg(OH)2
Solubility: Dilute Acids
Synonyms: Milk of Magnesia, Magnesium Hydrate
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...