Magnesium Chloride Hexahydrate

Supplier: ALDON CORP SE

MM0050-100G MM0050-500G MM0050-100GR MM0050-500GR MM0062-CONC MM0062-500ML MM0061-500ML
470045-526EA 18.4 CAD
470045-526 470301-407 470301-632 470301-634 470301-636 470301-638 470225-660
Magnesium Chloride Hexahydrate
Chemicals for Science Education

CAS Number: 7791-18-6
Formula: MgCl2 6H2O
Density: 1.57 g/mL
Solubility: Water and Alcohol
Synonyms: Magnesium Chloride 6-Hydrate
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR