Magnesium Carbonate

Supplier: ALDON CORP SE

MM0035-56G MM0040-500G MM0040-100G
470301-622EA 15.8 CAD
470301-622 470301-624 470301-626
Magnesium Carbonate
Chemicals for Science Education

CAS Number: 39409-82-0
Formula: 4MgCO3·2Mg(OH)2·4H2O/5H2O
Density (g/mL): 2.16
Solubility: Acids and Slightly in Water
Synonyms: Magnesium Carbonate basic, tetrahydrate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR