Magnesium Bromide Hexahydrate

Supplier: ALDON CORP SE

MM0190-500G MM0024-500ML
470301-620EA 65.55 CAD
470301-620 470225-780
Magnesium Bromide Hexahydrate
Chemicals for Science Education

CAS Number: 13446-53-2
Formula Weight: 292.25
Formula: MgBr2·6H2O
Synonyms: Magnesium Bromide 6-Hydrate
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR