Magnesium

Supplier: ALDON CORP SE

MM0022-100G MM0010-25G MM010-SS/2 MM0022-500G MM0022-100GR MM0022-500GR
470045-575EA 108.4 CAD
470045-575 470301-415 470301-416 470301-652 470301-654 470301-656
Magnesium
Chemicals for Science Education

CAS Number: 7439-95-4
Formula: Mg
Solubility: Acids
Shelf Life: 36 months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This item is flammable.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR