Prepared Luria Broth Media
Supplier: Avantor
470180-652PK 45.75 CAD
470180-652 470180-720
Prepared Luria Broth Media
Media for Microbiology Dehydrated Media for Microbiology
This high-nutrient broth is a great general purpose media. Especially useful for growing E. coli cultures
  • Sterile, pre-prepared media; just inoculate and incubate
Each tube contains 9mL of liquid broth.
Each bottle contains 200mL of liquid broth.