Lung Volume Bag Sets
Supplier: NASCO
SB06915 SB51114 SB51112
470008-080KT 93.55 CAD
470008-080 470008-082 470008-084
Lung Volume Bag Sets
Respirators Respiratory Systems
This set is a simple and inexpensive tool that measures vital capacity.
  • Set measures vital capacity
  • Set also compares effects of height, age, smoking, and more
  • Four mouthpieces included
  • Simple
  • Inexpensive
Vital capacity is the volume of air a person can expel after taking the deepest possible breath.
This set can also be used to compare effects of height, age, physical activity, and smoking.
Ordering information: This kit includes four holders, four mouthpieces, and four heavy-gauge plastic bags calibrated to 6 L.