Boyle's Law Apparatus
Supplier: VWR International
P12030 6471001
470148-928CS 1431.15 CAD
470148-928 470006-358
Boyle's Law Apparatus
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
This apparatus makes it easy for students to learn about Boyle's Law.
  • Calibrated Plastic Syringe
  • Wooden Base
  • Lubricated Piston
  • Various Sizes of Weights
  • Students Plot Pressure against the Volume of Gases in the Cylinder
The calibrated plastic syringe is anchored onto a wooden base.
A lubricated piston is mounted to a platform that holds various size of weights, so students can plot pressure against the volume of the gases in the cylinder.