Ward's® Plasmid DNA for Transformations
Supplier: Edvotek
851306B
470177-184EA 44.15 CAD
470177-184 470184-902
Ward's® Plasmid DNA for Transformations
Vectors, Plasmids and Libraries
These plasmids allow you to perform bacterial transformations, conferring new antibiotic resistance, as a visual way of demonstrating gene transfer.
  • Purchase only the amount of plasmid you need
  • Many plasmids already contain loading dye for added convenience
Plasmid pUC19: 2686 base pairs. Confers antibiotic resistance to ampicillin and complements defects in b-galactosidase in appropriate bacterial strains. Isolated from E. coli JM101.
pBR322: 4361 base pairs. Commonly used plasmid cloning vector for E. coli, and confers resistance to Ampicillin and Tetracycline.
Ordering information: These products are perishable materials, and are shipped next-day air on ice to ensure freshness. Please specify a delivery date at order time.