Litmus

Supplier: ALDON CORP SE

LL0230-25G LL0240-500ML_U
470301-602EA 42.35 CAD
470301-602 470301-604
Litmus
Chemicals for Science Education

CAS Number: 1393-92-6
Formula Weight: Approximately 3300
Solubility: Water
Synonyms: 7-Hydroxy-2-Phenoxazone
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR