Liquid Convection Apparatus
Supplier: VWR International
P20318
470148-912CS 4083.85 CAD
470148-912
Liquid Convection Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Effective Method of Showing Normally Invisible Currents
A rectangular glass tube has an opening at the top in which to pour water and introduce dye, which is available separately. When a corner of the tube is heated, the current is visible in the movement of the dye.