Limewater

Supplier: ALDON CORP SE

LL0170-1L LL0170-3.8L LL0170-500ML
470301-373EA 23.15 CAD
470301-373 470301-374 470301-375
Limewater
Chemicals for Science Education

CAS Number: 1305-62-0
Formula: Ca(OH)2
Formula Weight: 74.10
Shelf Life (months): 12
Storage: Green
Synonyms: Calcium Hydroxide Solution

Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR