Lens Sets
Supplier: Wards
PH0538A/CB PH0538B/CB PH0538D PH0538C
470018-264KT 27.75 CAD
470018-264 470014-446 470135-012 470135-014
Lens Sets
Educational Materials Physics Educational Materials Optics Learning Activities
Choose high quality crown glass or shatter-resistant acrylic lenses.
  • Wide range of lens sizes and materials
  • 6 lenses: double convex, plano-convex, concavo-convex, double concave, plano-concave, and convexo-concave
Perfect for optical benches or general use, these sets contain one each of the six lens types shown. Glass lenses resist scratching, but are more brittle than acrylic. Sets come in a cardboard storage box. Acrylic sets include an activity guide. Available in 38 mm or 50 mm diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable