Lens and Mirror Support
Supplier: ELECTRO MECHANICAL INSTRUMENTS
6008 6006
470006-932EA 7.55 CAD
470006-932 470006-934
Lens and Mirror Support
Mirrors
These simple supports are used to hold lenses and circular mirrors on a meter stick optical bench.
  • Available in two diameters, 38 and 75 mm
Fancy optics benches are very expensive, but a simple meter stick can give accurate results for most student laboratory applications. These supports will hold your lenses or mirrors securely in line, while making focal point measurements clear and easy.