Leakproof Dispensing Bottles with Spout Cap
Supplier: Thermo Fisher Scientific
2411-0015 2411-0060 2411-0125 2411-0250
470211-782CS 551.42 CAD
470211-782 470211-784 470211-786 470211-788
Leakproof Dispensing Bottles with Spout Cap
Bottles Dropping Bottles
No spill dropper.
  • LDPE bottle construction
  • Attached polypropylene caps included
  • Delivers just one drop at a time with a convenient one-hand operation
  • Captive spout cap stays in place when bottle is squeezed
The attached spout caps won’t pop off when bottle is squeezed, so they can’t get lost.