Lead(II) Nitrate

Supplier: ALDON CORP SE

LL0124-500G LL0125-100GR LL0125-500GR LL0125-CAP LL0132-CONC LL0133-50ML LL0133-500ML LL0132-500ML LL0124-100G LL0124-250G LL0131-500ML
470301-358EA 245.95 CAD
470301-358 470301-359 470301-360 470301-518 470301-520 470301-522 470301-524 470301-526 470301-528 470301-530 470225-666
Lead(II) Nitrate
Chemicals for Science Education

CAS Number: 10099-74-8
Formula: PbNO32
Density: 4.53 g/mL
Solubility: Water and Alcohol
Synonyms: Lead Dinitrate
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is toxic, an oxidizer, and flammable.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR