Lead(II) Acetate Trihydrate

Supplier: ALDON CORP SE

LL0090-500G LL0093-500ML LL0095-500ML
470301-510EA 48.8 CAD
470301-510 470225-710 470225-738
Lead(II) Acetate Trihydrate
Chemicals for Science Education

CAS Number: 6080-56-4
Formula Weight: 379.34
Formula: Pb(CH3COO)2·3H2O
Hazard Info: Toxic, Irritant, Carcinogen
Density (g/mL): 2.55
Boiling Point (°C): 280
Freezing Point (°C): 75
Solubility: Water and Glycerin
Synonyms: Lead Acetate
Shelf Life (months): 12
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR