Lead

Supplier: ALDON CORP SE

LL0085-500G LL0077-100G LL0081-476G LL0082-503G LL0086-500G LL0080-500G LL0077-500G LL0070-100G LL0081-1SHT
470301-369EA 96.6 CAD
470301-369 470301-538 470301-544 470301-546 470301-548 470301-550 470301-896 470301-900 470229-734
Lead
Chemicals for Science Education

CAS Number: 7439-92-1
Formula Weight: 207.19
Formula: Pb
Hazard Info: Toxic
Density (g/mL): 11.34
Boiling Point (°C): 1753
Freezing Point (°C): 327
Solubility: Dilute Nitric Acid
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR