Latex Disposable Gloves
Supplier: Dival Safety & Supplies
R10UN-SM R10UN-LG 5055SM 5055LG R10UN-MD 5055XL
470152-192BX 40.15 CAD
470152-192 470152-194 470157-558 470157-562 470195-112 470356-762
Latex Disposable Gloves
Gloves
With excellent tactile sensitive, these wrist-length gloves are ambidextrous and disposable.
  • Ambidextrous
  • Non sterile
  • Economy hand protection
  • Wrist-length
  • Aligned with GLOBE protocols
Perfect for student use, they feature a textured finish for sure grip and a beaded cuff to keep them in place.
Latex gloves reduce costs for hand protection when glove usage is high. Pre-powered option available for easy removal in your choice of sizes.