Latex Injection Medium

Supplier: ALDON CORP SE

LL0058-1L LL0058-3.8 LL0057-1L LL0057-3.8 LL0059-1L LL0060-1L LL0060-500ML
470024-608EA 121.1 CAD
470024-608 470024-610 470024-612 470024-614 470024-616 470301-502 470301-824
Latex Injection Medium
Chemicals for Science Education

Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 0.95
Boiling Point (°C): 100
Solubility: Organic Solvents
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR