Premium Rock Collection

Supplier: GEOSCIENCE INDUSTRIES INC SE
1750-000
470151-166EA 130.75 CAD
470151-166
Premium Rock Collection
Specimens Rock Specimens Rock Collections
The Premium Rock Collection features 12 palm-sized specimens representing all three rock types: igneous, metamorphic, and sedimentary.

  • Twelve Palm-Sized Specimens
  • All Three Rock Types Represented
  • All Specimens Are Numbered and Keyed
  • Length: 2"
  • Width: 3"

Perfectly sized for identifying details in the rock, each specimen is numbered and keyed to an ID sheet that features additional data.

Specimens are approximately 2 x 3."

Ordering information: This kit includes classification charts, a rock cycle chart, and a magnifier.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR