LaMotte® Smart3 Colorimeter and Reagents

Supplier: LaMotte
1910 3650-SC 3700-01-SC
470002-194EA 2334.2 CAD
470002-194 470349-454 470349-452
LaMotte® Smart3 Colorimeter and Reagents
Colorimeters Multiparameter Colorimeters
This direct-reading colorimeter is ideal for complete, on-site analysis.

  • Weight: 11 oz.
  • Transmittance Range: 0 to 125%
  • Resolution: 1%

Any of 75 preprogrammed tests and up to 20 user-entered tests for reagent systems can be selected quickly and easily. Results are displayed in concentration (ppm) or absorbance. The data logger holds up to 500 time and date-stamped data points. It also features self-diagnostics with error/warning messages, RS232 port, auto zero, and auto shutoff. Super Twist LCD display enhances readability, and optional software provides flexible data storage and manipulation. Transmittance range is 0-125%, resolution is 1%, accuracy is ±2% fs, absorbance range is 0-2.0 AU, resolution is 0.001 AU, and accuracy is ±0.005 AU.

Only ACS raw materials are used to manufacture these reagent systems for the Smart Colorimeter.

Accessories information: Additional factors are available; call for a full list.

Ordering information: An AC adapter, 9 V battery, and four sample tubes are included, along with an instruction manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR