LaMotte® pH TRACER
Supplier: LaMotte
1741 1733
470012-552EA 279.2 CAD
470012-552 470012-554
LaMotte® pH TRACER
Electrochemistry Meters pH and Multi-Parameter Meters
Store Up to 15 Readings
  • Automatic Temperature Compensation
  • Auto-off to Conserve Battery Life
  • Rugged Flat Electrode
For clear readings everytime, this pH meter features a large, bold display, which shows pH and temperature of a sample in numbers or bar graphs. A built-in memory stores up to 15 readings. Automatic shut-off and low battery indicator are additional features. It includes 4, 7, and 10 pH buffer tablets and four LR-44 batteries. pH range: 0–14, resolution: 0.01 pH, accuracy: ±0.01 pH. Temperature range: 32–149°F (0–65°C), resolution: 0.1°F/C.
pH Range: 0 - 14. Resolution: 0.01 Accuracy: ±0.01 pH.
Temperature Range: 32–149 °F; 0–65 °C. Resolution: 0.1°F/°C.