LaMotte® NPK Soil Test Kit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5880-6 1382
470157-410CS 385.55 CAD
470157-410 470222-296
LaMotte® NPK Soil Test Kit
Standards Environmental Analysis Standards
Safe, easy, economical test for three factors.
  • No hazardous compounds required for testing
  • Useful for the lab and field work
  • Includes instructions
  • Aligned with GLOBE protocols
Test for nitrogen, potassium, and phosphorus with one kit; the special TesTabs, nonhazardous reagents in tablet form, make it simple. The kit comes with testing apparatus, illustrated instructions, and a laminated color chart for comparison all packaged in a light carrying case for transportation to the field. You get enough materials for 50 tests each.