LaMotte® Acidity Test Kit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5024 5024
470005-816EA 55.8 CAD
470005-816 470201-278
LaMotte® Acidity Test Kit
Standards Environmental Analysis Standards
Accurate pH Testing
  • 100 Tests
  • Spot Testing of Your Soil
Tests are made with a wide range indicator and give the pH of soil to ±0.5 pH units. Includes 50 tests, chemical indicator, porcelain plates, color comparator chart, complete Instructions, pocket-size case and a soil test handbook.