Lambda Markers
Supplier: Avantor
470175-926EA
64.7
CAD
470175-926
470175-928
470175-930
470175-932
Lambda Markers
Electrophoresis Reagents
Molecular Weight Markers
These DNA markers are used to determine size of unknown DNA fragments. If using within 2–3 days, they do not need to be refrigerated and can be stored right on a shelf. If storing for longer time periods, freezing is recommended.
Ship date is required.