Ward's® Live Lactobacillus casei Culture
Supplier: Avantor
470179-124EA 31.5 CAD
470179-124 470179-524
Ward's® Live Lactobacillus casei Culture
Specimens Live Specimens Bacteria - Living Material
Isolated from milk and cheese products. Can be used to ferment milk. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Rods in chains
  • Gram Stain: Positive
  • Respiration: Facultatively anaerobic, sometimes microaerophilic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This product is for educational use only, do not use this organism to ferment milk that will be consumed.
Though this organism is not pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.