Ward's® Labradorite (Cleavage)
Supplier: Avantor
470025-658PK 66.5 CAD
470025-658 470025-660 470025-662
Ward's® Labradorite (Cleavage)
Specimens Mineral Specimens Classroom Minerals
Feldspar group; good cleavages of pure soda lime feldspar; most show twinning and labradorescence.
  • WARD’S offers different specimen sizes to meet your various needs
  • Student specimens, hand specimens, and bulk packs are available