Laboratory Tote Trays
Supplier: SHIRLEY KS STORAGE TRAYS
T010001 T013501 T013301
470005-902EA 47.7 CAD
470005-902 470149-748 470149-776
Laboratory Tote Trays
Trays/Dishes/Pots Laboratory Trays/Dishes
These sturdy trays are a useful item to have in every laboratory.
  • They will not chip, crack, peel, or break under normal conditions
  • Smooth inner surface and rounded corners assure easy cleaning
  • Each has a recessed front with a convenient label holder
The trays are acid and alkali resistant.